Learn More
Medchemexpress LLC HY-50686 500mg , Tivantinib ARQ 197 CAS:905854-02-6 Purity:98%
Supplier: Medchemexpress LLC HY50686500MG
Medchemexpress, HY-50686 500mg Tivantinib ARQ 197 CAS:905854-02-6 Formula:C23H19N3O2 Ki: 355 nM (c-Met) Purity:98% Tivantinib is a novel and highly selective c-Met tyrosine kinase inhibitor with K i of 355 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.