Learn More
Selleck Chemical LLC Tivantinib S2753-50mg
SDP
Catalog No. 503111837
Encompass_Preferred
Tivantinib is the first non-ATP-competitive c-Met inhibitor with Ki of 0 355 M in a cell-free assay little activity to Ron and no inhibition to EGFR InsR PDGFR or FGFR1/4 Tivantinib (ARQ 197) induces a G2/M arrest and apoptosis
Catalog No. 50-311-1837 Supplier Selleck Chemical LLC Supplier No. S275350MG
Tivantinib is the first non-ATP-competitive c-Met inhibitor with Ki of 0 355 M in a cell-free assay little activity to Ron and no inhibition to EGFR InsR PDGFR or FGFR1/4 Tivantinib (ARQ 197) induces a G2/M arrest and apoptosis