Learn More
Selleck Chemical LLC Tivantinib (ARQ 197) 10mg 905854-02-6
SDP
Catalog No. 501362904
Encompass_Preferred
Tivantinib (ARQ 197) is the first non-ATP-competitive c-Met inhibitor with Ki of 0.355 ?M in a cell-free assay, little activity to Ron, and no inhibition to EGFR, InsR, PDGFR? or FGFR1/4. Tivantinib (ARQ 197) induces a G2/M arrest and apoptosis. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2904 Supplier Selleck Chemical LLC Supplier No. S275310MG
Tivantinib (ARQ 197) is the first non-ATP-competitive c-Met inhibitor with Ki of 0.355 ?M in a cell-free assay, little activity to Ron, and no inhibition to EGFR, InsR, PDGFR? or FGFR1/4. Tivantinib (ARQ 197) induces a G2/M arrest and apoptosis. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.