Learn More
Medchemexpress LLC Tiviciclovir hydrochloride | 00-00-0 | 275.69 g/mol | C9H14ClN5O3 | 100 MG
SDP
Catalog No. 5000243841
Encompass_Preferred
Tiviciclovir hydrochloride is an antiviral guanosine analog that inhibits hepatitis B virus replication. Provided for research use only, the compound is supplied as the hydrochloride salt with formula C9H14ClN5O3 and a molecular weight of 275.69 g/mol. Solubility and storage recommendations follow the supplier datasheet.
  • Antiviral guanosine analog with HBV inhibitory activity.
  • Suitable for in vitro research applications (research use only).
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic treatment.
  • Stable as a solid when stored sealed at 4°C.
  • In solution, store at -80°C for up to 6 months or -20°C for up to 1 month.
  • Includes molecular formula and molecular weight for reference.
Catalog No. 50-002-43841 Supplier Medchemexpress LLC Supplier No. HY106233B100MG
Tiviciclovir hydrochloride is an antiviral guanosine analog that inhibits hepatitis B virus replication. Provided for research use only, the compound is supplied as the hydrochloride salt with formula C9H14ClN5O3 and a molecular weight of 275.69 g/mol. Solubility and storage recommendations follow the supplier datasheet.
  • Antiviral guanosine analog with HBV inhibitory activity.
  • Suitable for in vitro research applications (research use only).
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic treatment.
  • Stable as a solid when stored sealed at 4°C.
  • In solution, store at -80°C for up to 6 months or -20°C for up to 1 month.
  • Includes molecular formula and molecular weight for reference.