Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tiviciclovir hydrochloride | 00-00-0 | 99.4% | 275.69 g/mol | C9H14ClN5O3 | 5 MG
SDP

Catalog No. 5000243167
Encompass_Preferred

Tiviciclovir hydrochloride is an antiviral guanosine analog used as a hepatitis B virus (HBV) inhibitor in preclinical research. It is supplied as the hydrochloride salt, with molecular formula C9H14ClN5O3 and molecular weight 275.69 g/mol. The product listing and datasheet include solubility, formulation guidance, storage recommendations, and batch-specific purity information for laboratory use.

  • Has antiviral activity against hepatitis B virus.
  • Provided as a hydrochloride salt to aid solubility.
  • Soluble in DMSO at 100 mg/mL; ultrasonic aid recommended.
  • Formulation guidance for in vivo dosing is included.
  • Storage recommendations for dry material and solutions are provided.
  • Includes batch-specific certificate of analysis indicating high purity.

Catalog No. 50-002-43167 Supplier Medchemexpress LLC Supplier No. HY106233B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tiviciclovir hydrochloride is an antiviral guanosine analog used as a hepatitis B virus (HBV) inhibitor in preclinical research. It is supplied as the hydrochloride salt, with molecular formula C9H14ClN5O3 and molecular weight 275.69 g/mol. The product listing and datasheet include solubility, formulation guidance, storage recommendations, and batch-specific purity information for laboratory use.

  • Has antiviral activity against hepatitis B virus.
  • Provided as a hydrochloride salt to aid solubility.
  • Soluble in DMSO at 100 mg/mL; ultrasonic aid recommended.
  • Formulation guidance for in vivo dosing is included.
  • Storage recommendations for dry material and solutions are provided.
  • Includes batch-specific certificate of analysis indicating high purity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.