Learn More
Medchemexpress LLC Tivozanib | 475108-18-0 | 99.8% | 454.86 | 100 MG
SDP
Catalog No. 5000344959
Encompass_Preferred
Tivozanib is a selective, orally active inhibitor for vascular endothelial growth factor receptor (VEGFR)-1, 2, and 3. It demonstrates antitumor efficacy and is suitable for research use.
  • Selectively inhibits VEGFR-1, 2, and 3 with low nanomolar IC50 values
  • Inhibits VEGF-induced proliferation and migration of HUVECs
  • Exhibits antitumor efficacy across various cancer models in athymic mice
  • Reversibly suppresses vascular permeability and angiogenesis
Catalog No. 50-003-44959 Supplier Medchemexpress LLC Supplier No. HY10977200MG
Tivozanib is a selective, orally active inhibitor for vascular endothelial growth factor receptor (VEGFR)-1, 2, and 3. It demonstrates antitumor efficacy and is suitable for research use.
  • Selectively inhibits VEGFR-1, 2, and 3 with low nanomolar IC50 values
  • Inhibits VEGF-induced proliferation and migration of HUVECs
  • Exhibits antitumor efficacy across various cancer models in athymic mice
  • Reversibly suppresses vascular permeability and angiogenesis