Learn More
Apexbio Technology LLC Tivozanib (AV-951), 10mg. Cas: 475108-18-0 MFCD: MFCD15146788. VEGFR inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A225110
Encompass_Preferred
Tivozanib is an inhibitor of tyrosine kinase with IC50 value of 160 pmol/L against VEGFR-2 [1].Tivozanib is a quinoline-urea derivative. As a 2nd generation TKI, it has picomolar potency against VEGFR-1, -2 and -3, and minimal c-kit inhibition. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4739