Learn More
Apexbio Technology LLC Tivozanib (AV-951) 475108-18-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064114
Encompass_Preferred
Tivozanib (AV-951 CAS 475108-18-0) is a quinoline-urea derivative functioning as a potent inhibitor of vascular endothelial growth factor receptors (VEGFR) It primarily targets VEGFR-2 kinase demonstrating significant inhibitory activity in picomolar ranges (IC50 of approximately 160 pM) Additionally Tivozanib displays considerable selectivity causing minimal off-target interaction with c-kit In cellular assays it inhibits PDGFR and c-kit phosphorylation at nanomolar levels Preclinical evaluations revealed antitumor activity across multiple solid tumor xenograft models particularly renal cell carcinoma (RCC) Its mechanism and selectivity profile render it valuable for cancer research clinical investigation and preclinical efficacy assessments
Catalog No. 50-000-64114 Supplier Apexbio Technology LLC Supplier No. A22515.1
Tivozanib (AV-951 CAS 475108-18-0) is a quinoline-urea derivative functioning as a potent inhibitor of vascular endothelial growth factor receptors (VEGFR) It primarily targets VEGFR-2 kinase demonstrating significant inhibitory activity in picomolar ranges (IC50 of approximately 160 pM) Additionally Tivozanib displays considerable selectivity causing minimal off-target interaction with c-kit In cellular assays it inhibits PDGFR and c-kit phosphorylation at nanomolar levels Preclinical evaluations revealed antitumor activity across multiple solid tumor xenograft models particularly renal cell carcinoma (RCC) Its mechanism and selectivity profile render it valuable for cancer research clinical investigation and preclinical efficacy assessments