Learn More
Selleck Chemical LLC Tivozanib (AV-951) 50mg 475108-18-0 KRN-951
SDP
Catalog No. 508155
Encompass_Preferred
Tivozanib (AV-951, KRN-951) is a potent and selective VEGFR inhibitor for VEGFR1/2/3 with IC50 of 30 nM/6.5 nM/15 nM, and also inhibits PDGFR and c-Kit, low activity observed against FGFR-1, Flt3, c-Met, EGFR and IGF-1R. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-815-5 Supplier Selleck Chemical LLC Supplier No. S120750MG
Tivozanib (AV-951, KRN-951) is a potent and selective VEGFR inhibitor for VEGFR1/2/3 with IC50 of 30 nM/6.5 nM/15 nM, and also inhibits PDGFR and c-Kit, low activity observed against FGFR-1, Flt3, c-Met, EGFR and IGF-1R. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.