Learn More
Medchemexpress LLC Tivozanib hydrochloride hydrate | 682745-41-1 | 99.8% | 509.34 | 10 MG
SDP
Catalog No. 5000344826
Encompass_Preferred
Tivozanib hydrochloride hydrate is the hydrate hydrochloride form of Tivozanib. It is a selective, orally active inhibitor for vascular endothelial growth factor receptor (VEGFR)-1, 2, and 3, with IC50s of 30, 6.5, and 15 nM, respectively. It exhibits antitumor efficacy.
  • Selective, orally active inhibitor for VEGFR-1, 2, and 3.
  • Exhibits antitumor efficacy.
  • Inhibits the phosphorylation of VEGFR-1, VEGFR-2, and VEGFR-3.
  • Inhibits VEGF-induced proliferation and migration of HUVECs.
  • Selectively inhibits VEGF-stimulated phosphorylation of MAPKs in endothelial cells.
  • Exhibits antitumor efficacy against various cancers in athymic mice models.
  • Reversibly suppresses vascular permeability and angiogenesis.
Catalog No. 50-003-44826 Supplier Medchemexpress LLC Supplier No. HY10977A100MG
Tivozanib hydrochloride hydrate is the hydrate hydrochloride form of Tivozanib. It is a selective, orally active inhibitor for vascular endothelial growth factor receptor (VEGFR)-1, 2, and 3, with IC50s of 30, 6.5, and 15 nM, respectively. It exhibits antitumor efficacy.
  • Selective, orally active inhibitor for VEGFR-1, 2, and 3.
  • Exhibits antitumor efficacy.
  • Inhibits the phosphorylation of VEGFR-1, VEGFR-2, and VEGFR-3.
  • Inhibits VEGF-induced proliferation and migration of HUVECs.
  • Selectively inhibits VEGF-stimulated phosphorylation of MAPKs in endothelial cells.
  • Exhibits antitumor efficacy against various cancers in athymic mice models.
  • Reversibly suppresses vascular permeability and angiogenesis.