Learn More
Medchemexpress LLC Tivozanib hydrochloride hydrate | 682745-41-1 | 99.8% | 509.34 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY10977A10MG
Encompass_Preferred
Tivozanib hydrochloride hydrate is the hydrate hydrochloride form of Tivozanib. It is a selective, orally active inhibitor for vascular endothelial growth factor receptor (VEGFR)-1, 2, and 3. This compound exhibits antitumor efficacy.
  • Selective, orally active inhibitor for VEGFR-1, 2, and 3
  • Exhibits antitumor efficacy
  • Available in solid form, white to off-white color
  • Soluble in DMSO (125 mg/mL)
  • Inhibits the phosphorylation of VEGFR-1, VEGFR-2, and VEGFR-3
  • Inhibits VEGF-induced proliferation and migration of HUVECs
  • Selectively inhibits VEGF-stimulated phosphorylation of MAPKs in endothelial cells
  • Exhibits antitumor efficacy against various cancers in athymic mice models
  • Reversibly suppresses vascular permeability and angiogenesis in tumor-bearing rats
Catalog No. 50-003-44268