Learn More
Medchemexpress LLC Tivozanib hydrochloride hydrate | 682745-41-1 | 99.8% | 509.34 | 50 MG
SDP
Catalog No. 5000344632
Encompass_Preferred
Tivozanib hydrochloride hydrate is the hydrate hydrochloride form of Tivozanib. It is a selective, orally active inhibitor for vascular endothelial growth factor receptor (VEGFR)-1, 2, and 3, with IC50s of 30, 6.5, and 15 nM, respectively. It demonstrates antitumor efficacy.
  • Selective, orally active inhibitor for VEGFR-1, 2, and 3
  • Exhibits antitumor efficacy
  • Inhibits phosphorylation of VEGFR-1, VEGFR-2, and VEGFR-3 with IC50s of 0.16-0.24 nM in vitro
  • Inhibits VEGF-induced proliferation and migration of HUVECs in vitro
  • Selectively inhibits VEGF-stimulated phosphorylation of MAPKs in endothelial cells in vitro
  • Shows antitumor efficacy against breast, colon, hepatic, lung, ovarian, pancreatic, and prostate cancer in athymic mice models in vivo
  • Reversibly suppresses vascular permeability and angiogenesis in Calu-6 tumor bearing rats in vivo
Catalog No. 50-003-44632 Supplier Medchemexpress LLC Supplier No. HY10977A10MM/1ML
Tivozanib hydrochloride hydrate is the hydrate hydrochloride form of Tivozanib. It is a selective, orally active inhibitor for vascular endothelial growth factor receptor (VEGFR)-1, 2, and 3, with IC50s of 30, 6.5, and 15 nM, respectively. It demonstrates antitumor efficacy.
  • Selective, orally active inhibitor for VEGFR-1, 2, and 3
  • Exhibits antitumor efficacy
  • Inhibits phosphorylation of VEGFR-1, VEGFR-2, and VEGFR-3 with IC50s of 0.16-0.24 nM in vitro
  • Inhibits VEGF-induced proliferation and migration of HUVECs in vitro
  • Selectively inhibits VEGF-stimulated phosphorylation of MAPKs in endothelial cells in vitro
  • Shows antitumor efficacy against breast, colon, hepatic, lung, ovarian, pancreatic, and prostate cancer in athymic mice models in vivo
  • Reversibly suppresses vascular permeability and angiogenesis in Calu-6 tumor bearing rats in vivo