Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tivozanib (hydrochloride hydrate) | 682745-41-1 | MFCD34184484 | >99.0% | 509.3 g/mol | C22H22Cl2N4O6 | 5 MG
SDP

Catalog No. 5000263523
Encompass_Preferred

Tivozanib (hydrochloride hydrate) is the monohydrochloride monohydrate salt of tivozanib, a selective, orally active inhibitor of vascular endothelial growth factor receptors (VEGFRs). It is supplied for research and analytical applications, including in vitro biochemical and cellular assays.

  • Selective VEGFR inhibitor suitable for target validation and signaling studies.
  • Hydrochloride hydrate solid form for improved handling and stability.
  • Reported high purity (>99%) from supplier analyses.
  • Molecular formula C22H22Cl2N4O6; molecular weight 509.3 g/mol.
  • Provided in small research quantities for assay development and SAR work.

Catalog No. 50-002-63523 Supplier Medchemexpress LLC Supplier No. HY10977A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tivozanib (hydrochloride hydrate) is the monohydrochloride monohydrate salt of tivozanib, a selective, orally active inhibitor of vascular endothelial growth factor receptors (VEGFRs). It is supplied for research and analytical applications, including in vitro biochemical and cellular assays.

  • Selective VEGFR inhibitor suitable for target validation and signaling studies.
  • Hydrochloride hydrate solid form for improved handling and stability.
  • Reported high purity (>99%) from supplier analyses.
  • Molecular formula C22H22Cl2N4O6; molecular weight 509.3 g/mol.
  • Provided in small research quantities for assay development and SAR work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.