Learn More
Cayman Chemical TIvoznIbhydrate 10mg
SDP
Catalog No. 502980186
Encompass_Preferred
An orally available, selective VEGFR inhibitor (IC50s = 0.21, 0.16, and 0.24 nM for VEGFR1, VEGFR2, and VEGFR3, respectively); also inhibits c-Kit and PDGFRβ (IC50s = 1.63 and 1.72 nM, respectively)
Catalog No. 50-298-0186 Supplier Cayman Chemical Supplier No. 1849310 MG
An orally available, selective VEGFR inhibitor (IC50s = 0.21, 0.16, and 0.24 nM for VEGFR1, VEGFR2, and VEGFR3, respectively); also inhibits c-Kit and PDGFRβ (IC50s = 1.63 and 1.72 nM, respectively)