Learn More
Medchemexpress LLC Tixocortol pivalate | 55560-96-8 | 96.8% | C26H38O5S | 10MG
SDP
Catalog No. 5000260028
Encompass_Preferred
Tixocortol pivalate is a synthetic corticosteroid used in research as a topical anti-inflammatory agent and diagnostic allergen. It exhibits limited systemic glucocorticoid and mineralocorticoid activity and is supplied as a purified solid for laboratory applications.
  • Topical anti-inflammatory corticosteroid.
  • Limited systemic glucocorticoid and mineralocorticoid activity.
  • Used as a diagnostic allergen in patch testing.
  • Provided as a purified solid for research applications.
  • Characterized by molecular formula C26H38O5S and molecular weight 462.64 g·mol⁻¹.
  • Purity typically around 96.8%.
Catalog No. 50-002-60028 Supplier Medchemexpress LLC Supplier No. HY13036510MG
Tixocortol pivalate is a synthetic corticosteroid used in research as a topical anti-inflammatory agent and diagnostic allergen. It exhibits limited systemic glucocorticoid and mineralocorticoid activity and is supplied as a purified solid for laboratory applications.
  • Topical anti-inflammatory corticosteroid.
  • Limited systemic glucocorticoid and mineralocorticoid activity.
  • Used as a diagnostic allergen in patch testing.
  • Provided as a purified solid for research applications.
  • Characterized by molecular formula C26H38O5S and molecular weight 462.64 g·mol⁻¹.
  • Purity typically around 96.8%.