Learn More
Medchemexpress LLC Tizanidine-d4 | 1188331-19-2 | 99.9% | 1 MG
SDP
Catalog No. 5000406232
Encompass_Preferred
Tizanidine-d4 is a deuterium labeled version of Tizanidine, a skeletal muscle relaxant and central α2-adrenoceptor agonist. It works by inhibiting the release of excitatory amino acids from spinal cord interneurons, reducing muscle spasticity. Tizanidine-d4 also shows anti-injury activity, can inhibit GI transport, and may inhibit proliferation, migration, and invasion of lung cancer cells by inducing cell apoptosis through specific signaling pathways. It is used to treat spasticity from conditions like multiple sclerosis, stroke, and spinal cord injury.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits anti-injury activity
  • Can inhibit gastrointestinal (GI) transport
  • May inhibit proliferation, migration, and invasion of lung cancer cells
  • Induces cell apoptosis by upregulating Nischarin and inhibiting AKT and Wnt3a/β-catenin signaling pathways
Catalog No. 50-004-06232 Supplier Medchemexpress LLC Supplier No. HYB0194S1MG
Tizanidine-d4 is a deuterium labeled version of Tizanidine, a skeletal muscle relaxant and central α2-adrenoceptor agonist. It works by inhibiting the release of excitatory amino acids from spinal cord interneurons, reducing muscle spasticity. Tizanidine-d4 also shows anti-injury activity, can inhibit GI transport, and may inhibit proliferation, migration, and invasion of lung cancer cells by inducing cell apoptosis through specific signaling pathways. It is used to treat spasticity from conditions like multiple sclerosis, stroke, and spinal cord injury.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits anti-injury activity
  • Can inhibit gastrointestinal (GI) transport
  • May inhibit proliferation, migration, and invasion of lung cancer cells
  • Induces cell apoptosis by upregulating Nischarin and inhibiting AKT and Wnt3a/β-catenin signaling pathways