Learn More
Medchemexpress LLC Tizanidine (hydrochloride) | 64461-82-1 | 99.93 % | 1 ML
SDP
Catalog No. 5000406640
Encompass_Preferred
Tizanidine hydrochloride is a skeletal muscle relaxant that acts as an orally effective central α2-adrenoceptor agonist. It primarily relaxes muscles by inhibiting the release of excitatory amino acids from spinal cord interneurons. Additionally, it exhibits anti-injury activity, can inhibit gastrointestinal (GI) transport, and has been shown to inhibit proliferation, migration, and invasion of lung cancer cells, inducing cell apoptosis. This compound is used to treat spasticity from conditions such as multiple sclerosis (MS), stroke, and spinal cord injury (SCI).
  • Skeletal muscle relaxant
  • Central α2-adrenoceptor agonist
  • Inhibits excitatory amino acid release
  • Exhibits anti-injury activity
  • Inhibits gastrointestinal (GI) transport
  • Inhibits lung cancer cell proliferation, migration, and invasion
  • Induces cell apoptosis
  • Treats spasticity from multiple sclerosis, stroke, and spinal cord injury
Catalog No. 50-004-06640 Supplier Medchemexpress LLC Supplier No. HYB0194A10MM/1ML
Tizanidine hydrochloride is a skeletal muscle relaxant that acts as an orally effective central α2-adrenoceptor agonist. It primarily relaxes muscles by inhibiting the release of excitatory amino acids from spinal cord interneurons. Additionally, it exhibits anti-injury activity, can inhibit gastrointestinal (GI) transport, and has been shown to inhibit proliferation, migration, and invasion of lung cancer cells, inducing cell apoptosis. This compound is used to treat spasticity from conditions such as multiple sclerosis (MS), stroke, and spinal cord injury (SCI).
  • Skeletal muscle relaxant
  • Central α2-adrenoceptor agonist
  • Inhibits excitatory amino acid release
  • Exhibits anti-injury activity
  • Inhibits gastrointestinal (GI) transport
  • Inhibits lung cancer cell proliferation, migration, and invasion
  • Induces cell apoptosis
  • Treats spasticity from multiple sclerosis, stroke, and spinal cord injury