Learn More
Cayman Chemical TIznIdIn-d4hydrochlorIde 500ug
SDP
Catalog No. 502935435
Encompass_Preferred
An internal standard for the quantification of tizanidine by GC- or LC-MS
Catalog No. 50-293-5435 Supplier Cayman Chemical Supplier No. 26521500 UG
An internal standard for the quantification of tizanidine by GC- or LC-MS