Learn More
Medchemexpress LLC Tizoxanide-d4 | 1246817-56-0 | 99.8% | 1 MG
SDP
Catalog No. 5000340886
Encompass_Preferred
Tizoxanide-d4 is the deuterium labeled form of Tizoxanide, which is the active metabolite of Nitazoxanide. This thiazolide anti-infective compound is effective against anaerobic bacteria, protozoa, and various viruses, including anti-HIV-1 activities. It is intended for research use only and can be used as a tracer or an internal standard for quantitative analysis.
  • Deuterium labeled compound
  • Active metabolite of Nitazoxanide
  • Effective against anaerobic bacteria, protozoa, and various viruses
  • Can be used as a tracer or internal standard
  • Useful for quantitative analysis by methods like NMR, GC-MS, or LC-MS
Catalog No. 50-003-40886 Supplier Medchemexpress LLC Supplier No. HY12687S1MG
Tizoxanide-d4 is the deuterium labeled form of Tizoxanide, which is the active metabolite of Nitazoxanide. This thiazolide anti-infective compound is effective against anaerobic bacteria, protozoa, and various viruses, including anti-HIV-1 activities. It is intended for research use only and can be used as a tracer or an internal standard for quantitative analysis.
  • Deuterium labeled compound
  • Active metabolite of Nitazoxanide
  • Effective against anaerobic bacteria, protozoa, and various viruses
  • Can be used as a tracer or internal standard
  • Useful for quantitative analysis by methods like NMR, GC-MS, or LC-MS