Learn More
Medchemexpress LLC Tizoxanide-d4 | 1246817-56-0 | 99.8% | 269.27 | C10H3D4N3O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY12687S5MG
Encompass_Preferred
Tizoxanide-d4 is the deuterium-labeled analogue of tizoxanide provided as a stable-isotope reference standard for research applications. It is intended for use as an internal standard in quantitative and metabolic studies where isotopic labeling improves accuracy and traceability. CAS: 1246817-56-0.
  • Deuterium-labeled analogue of tizoxanide.
  • Intended for research use only.
  • High chemical purity (≈99.8%).
  • Suitable as an internal standard for mass spectrometry.
  • Molecular formula C10H3D4N3O4S; molecular weight 269.27.
Catalog No. 50-002-64459