Learn More
Medchemexpress LLC HY-139397 5mg , TJ-M2010-5 CAS:1357471-57-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1393975MG
Encompass_Preferred
HY-139397 5mg TJ-M2010-5 CAS: 1357471-57-8 TJ-M2010-5 is a MyD88 inhibitor that binds to the TIR domain of MyD88 to interfere with its homodimerization, and the TLR/MyD88 signal pathway. TJ-M2010-5 can be used for the research of myocardial ischemia/reperfusion injury (MIRI). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9512