Learn More
Precision Glassblowing of Colorado TJA INJECTOR 1.0MM 12540701
SDP
Catalog No. 50420709
Encompass_Preferred
Quartz Injector, 1.0mm (2 red rings), for TJA 61E, Iris & Intrepid Radial, and for TJA 61E Trace, Iris AP, & Intrepid
Catalog No. 50-420-709 Supplier Precision Glassblowing of Colorado Supplier No. PG25040
Quartz Injector, 1.0mm (2 red rings), for TJA 61E, Iris & Intrepid Radial, and for TJA 61E Trace, Iris AP, & Intrepid