Learn More
Aviva Systems Biology Corp TJAP1 Antibody - C-terminal region (ARP42926_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42926_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TJAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TJAP1. Alternate names: DKFZp686F06131, PILT, TJP4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58365