Learn More
Aviva Systems Biology Corp TJP2 Antibody - middle region (ARP42941_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42941_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TJP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TJP2. Alternate names: MGC26306, X104, ZO-2, ZO2, DFNA51, DUP9q21.11, C9DUPq21.11ApplicationWB. Species reactivity: Dog, Horse, Human, Rabbit, Rat
Catalog No. 50-014-58369