Learn More
Medchemexpress LLC TK-OH | 861587-94-2 | 98.4% | 224.38 | 5 MG
SDP
Catalog No. 5000391089
Encompass_Preferred
TK-OH is a linker for LTB that can be used for cancer research. It is a viscous liquid, light yellow to yellow in color. The product is for research use only and not sold to patients.
  • Used for cancer research.
  • Viscous liquid appearance.
  • Light yellow to yellow color.
  • Soluble in DMSO at ≥ 175 mg/mL.
  • Pure form stable at -20°C for 3 years.
  • In solvent, stable at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-003-91089 Supplier Medchemexpress LLC Supplier No. HY1694355MG
TK-OH is a linker for LTB that can be used for cancer research. It is a viscous liquid, light yellow to yellow in color. The product is for research use only and not sold to patients.
  • Used for cancer research.
  • Viscous liquid appearance.
  • Light yellow to yellow color.
  • Soluble in DMSO at ≥ 175 mg/mL.
  • Pure form stable at -20°C for 3 years.
  • In solvent, stable at -80°C for 6 months or -20°C for 1 month.