Learn More
Aviva Systems Biology Corp Tk1 Antibody - middle region (ARP58541_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58541_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tk1. It was validated on Western Blot. . Alternate names: D530002A18Rik, Tk-1, Tk1a, Tk1bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-482