Learn More
ERA WATERS CORP TKN 1K STANDARD 500ML
SDP
Catalog No. NC9342153
Encompass
One 1,000 mg/L standard preserved with HCl in a poly bottle.
Catalog No. NC9342153 Supplier ERA WATERS CORP Supplier No. 996
One 1,000 mg/L standard preserved with HCl in a poly bottle.