Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TKN Digestion Reagent with Copper Sulfate, Reagents

Catalog No. 01333449
Change view
Click to view available options
Quantity:
4 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-449 4 L
01-333-450 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-449 Supplier Reagents Holdings Llc Supplier No. CT1007R04A
 Restricted Item
Add to Cart
Add to Cart

TKN Digestion Reagent with Copper Sulfate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Total Kjeldahl Nitrogen Digestion Reagent
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Blue
CAS 7778-80-5, 7664-93-9, 7732-18-5
Physical Form Liquid
Quantity 4 L
Packaging Poly Bottle
Grade Reagent
Chemical Name or Material TKN Digestion Reagent with Copper Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.