Learn More
Cole-Parmer Environmental Express TKN6012 Teardrop Stopper for TKN Digestion Tubes; 1/Ea
Catalog No. NC1739754 Shop All Cole-Parmer Products
Encompass
Teardrop stoppers are ideal for the capture and condesation of digestion fumes, preventing cross contamination.
Catalog No. NC1739754 Supplier Cole-Parmer Supplier No. 3520574
Teardrop stoppers are ideal for the capture and condesation of digestion fumes, preventing cross contamination.