Learn More
Aviva Systems Biology Corp TKT Antibody - N-terminal region (ARP48540_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48540_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TKT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TKT. Alternate names: FLJ34765, TKT1, TKApplicationWB, IHC. Species reactivity: Cow, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59853