Learn More
ABclonal Technology TKT/Transketolase
SDP
Catalog No. 5000011897
Encompass_Preferred
This gene encodes a thiamine-dependent enzyme which plays a role in the channeling of excess sugar phosphates to glycolysis in the pentose phosphate pathway Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene
Catalog No. 50-000-11897 Supplier ABclonal Technology Supplier No. A2714420UL
This gene encodes a thiamine-dependent enzyme which plays a role in the channeling of excess sugar phosphates to glycolysis in the pentose phosphate pathway Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene