Learn More
Aviva Systems Biology Corp TKTL1 Antibody - middle region (ARP53676_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53676_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TKTL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TKTL1. Alternate names: TKR, TKT2ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61086