Learn More
MSC Loctite Threadlocker, Red, High Strength, 250 mL
Catalog No. 501150309
Encompass
This high-strength, high-viscosity threadlocking adhesive is designed for the permanent locking and sealing of large bolts and studs. It cures in the absence of air between close-fitting metal surfaces, preventing loosening and leakage from shock and vibration. Suitable for fasteners up to 36 mm (1.5 in), it protects threads from rust and corrosion.
  • High strength, red liquid formulation
  • Permanent locking and sealing of large fasteners
  • Prevents loosening and leakage from shock and vibration
  • Suitable for bolts and studs up to 36 mm (1.5 in)
  • Disassembly possible with heat and hand tools
  • Meets MIL-S-46163A Type A Grade L
  • Operating temperature range up to 150 °C (300 °F)
  • Fluoresces for easy monitoring
Catalog No. 50-115-0309 Supplier MSC Supplier No. 90052853
This high-strength, high-viscosity threadlocking adhesive is designed for the permanent locking and sealing of large bolts and studs. It cures in the absence of air between close-fitting metal surfaces, preventing loosening and leakage from shock and vibration. Suitable for fasteners up to 36 mm (1.5 in), it protects threads from rust and corrosion.
  • High strength, red liquid formulation
  • Permanent locking and sealing of large fasteners
  • Prevents loosening and leakage from shock and vibration
  • Suitable for bolts and studs up to 36 mm (1.5 in)
  • Disassembly possible with heat and hand tools
  • Meets MIL-S-46163A Type A Grade L
  • Operating temperature range up to 150 °C (300 °F)
  • Fluoresces for easy monitoring