Learn More
Strem, An Ascensus Company CAS# 20398-06-5. 25g. Thallium(I) ethoxide, min. 95%. MFCD00009072
SDP
Supplier:  Strem, An Ascensus Company 93810525G
Encompass
CAS# 20398-06-5. 25g. Thallium(I) ethoxide, min. 95%. MFCD00009072. Molecular Weight: 249.43. Molecular Formula: Tl(OC2H5). Color/form: cloudy, dense liq. Strem# 93-8105. http://www.strem.com/catalog/v/93-8105/
Catalog No. 50-204-5434