Learn More
Perkin Elmer US LLC Thallium (Tl) Lumina Hollow Cathode Lamp
Catalog No. 509044262
Encompass_Preferred
Single Element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Thallium (Tl). Every genuine PerkinElmer lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime. Note: Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.
Catalog No. 50-904-4262 Supplier Perkin Elmer US LLC Supplier No. N3050183
Single Element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Thallium (Tl). Every genuine PerkinElmer lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime. Note: Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.