Learn More
Medchemexpress LLC TL13-112 | 2229037-19-6 | MFCD12922943 | 98.0% | 1002.57 | C49H60ClN9O10S | 10MM 1ML
SDP
Catalog No. 5000236667
Encompass_Preferred
TL13-112 is a proteolysis targeting chimera (PROTAC) designed to selectively induce degradation of anaplastic lymphoma kinase (ALK) in cellular models. It is supplied as a solid and as a 10 mM solution in DMSO for use in preclinical degradation studies, mechanistic assays, and target validation workflows.
  • Selective ALK degradation in cellular assays.
  • High purity suitable for biological studies.
  • Available as solid or 10 mM DMSO solution for convenient dosing.
  • Recommended storage conditions for powder and solvent preparations.
  • Applicable to mechanistic studies and preclinical target validation.
Catalog No. 50-002-36667 Supplier Medchemexpress LLC Supplier No. HY12391910MM 1ML
TL13-112 is a proteolysis targeting chimera (PROTAC) designed to selectively induce degradation of anaplastic lymphoma kinase (ALK) in cellular models. It is supplied as a solid and as a 10 mM solution in DMSO for use in preclinical degradation studies, mechanistic assays, and target validation workflows.
  • Selective ALK degradation in cellular assays.
  • High purity suitable for biological studies.
  • Available as solid or 10 mM DMSO solution for convenient dosing.
  • Recommended storage conditions for powder and solvent preparations.
  • Applicable to mechanistic studies and preclinical target validation.