Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TL13-112 | 2229037-19-6 | MFCD12922943 | 98.0% | 1002.57 | C49H60ClN9O10S | 50MG
SDP

Catalog No. 5000239133
Encompass_Preferred

TL13-112 is a small-molecule PROTAC designed to target and degrade anaplastic lymphoma kinase (ALK) in cellular models. It couples an ALK-binding warhead to a cereblon ligand to induce proteasomal degradation and has been characterized for potent ALK inhibition and cellular degradation activity in ALK-positive cancer cell lines.

  • Selective ALK PROTAC degrader that induces proteasomal degradation of ALK.
  • Potent ALK activity inhibition with IC50 = 0.14 nM.
  • Cellular ALK degradation with DC50 = 10 nM (H3122) and 40 nM (Karpas 299).
  • Also affects additional kinases including Aurora A, FER, PTK2, and RPS6KA1.
  • Molecular formula C49H60ClN9O10S and molecular weight ~1002.57 g/mol.
  • Purity reported as 98.01% by LC-MS.
  • Supplied in small milligram quantities for research use.

Catalog No. 50-002-39133 Supplier Medchemexpress LLC Supplier No. HY12391950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TL13-112 is a small-molecule PROTAC designed to target and degrade anaplastic lymphoma kinase (ALK) in cellular models. It couples an ALK-binding warhead to a cereblon ligand to induce proteasomal degradation and has been characterized for potent ALK inhibition and cellular degradation activity in ALK-positive cancer cell lines.

  • Selective ALK PROTAC degrader that induces proteasomal degradation of ALK.
  • Potent ALK activity inhibition with IC50 = 0.14 nM.
  • Cellular ALK degradation with DC50 = 10 nM (H3122) and 40 nM (Karpas 299).
  • Also affects additional kinases including Aurora A, FER, PTK2, and RPS6KA1.
  • Molecular formula C49H60ClN9O10S and molecular weight ~1002.57 g/mol.
  • Purity reported as 98.01% by LC-MS.
  • Supplied in small milligram quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.