Learn More
Selleck Chemical LLC TL13-112-E1846-1G
SDP
Catalog No. 503138974
Encompass_Preferred
TL13-112 is a degrader of anaplastic lymphoma kinase (ALK) with an IC50 of 0 60 nM It demonstrates anti-proliferative activity in non-small-cell lung cancer (NSCLC) and anaplastic large cell lymphoma (ALCL) cell lines
Catalog No. 50-313-8974 Supplier Selleck Chemical LLC Supplier No. E18461G
TL13-112 is a degrader of anaplastic lymphoma kinase (ALK) with an IC50 of 0 60 nM It demonstrates anti-proliferative activity in non-small-cell lung cancer (NSCLC) and anaplastic large cell lymphoma (ALCL) cell lines