Learn More
ACROBiosystems Cynomolgus/Rhesus macaque TL1A / TNFSF15 Protein, His Tag
Catalog No. 502331321
Encompass_Preferred
Cynomolgus/Rhesus macaque TL1A / TNFSF15, His Tag (TLA-C5241) is expressed from human 293 cells (HEK293). It contains AA Leu 72 - Leu 251 (Accession # F6S8F9-1).
Catalog No. 50-233-1321 Supplier ACROBiosystems Supplier No. TLAC52411MG
Cynomolgus/Rhesus macaque TL1A / TNFSF15, His Tag (TLA-C5241) is expressed from human 293 cells (HEK293). It contains AA Leu 72 - Leu 251 (Accession # F6S8F9-1).