Learn More
ACROBiosystems TL1A (Biotinylated) : DR3 Inhibitor Screening ELISA Kit, 96 tests
Catalog No. NC3874542
Encompass_Preferred
Competition ELISA kit for screening inhibitors that block human TL1A binding to human DR3. The kit supplies biotinylated TL1A, recombinant human DR3, a control neutralizing antibody, and all necessary reagents in a 96-well format for early-stage inhibitor screening and QC testing.
  • Competition ELISA format for inhibitor screening.
  • Includes biotinylated TL1A and recombinant DR3 protein.
  • Provides a positive control neutralizing antibody and assay reagents.
  • Designed for 96-well screening and QC workflows.
  • Intended species: human.
  • Store unopened at 2-8 °C and follow component storage after opening.
  • No sensitivity or dynamic range specified on the product page.
Catalog No. NC3874542 Supplier ACROBiosystems Supplier No. EP16896TESTS
Competition ELISA kit for screening inhibitors that block human TL1A binding to human DR3. The kit supplies biotinylated TL1A, recombinant human DR3, a control neutralizing antibody, and all necessary reagents in a 96-well format for early-stage inhibitor screening and QC testing.
  • Competition ELISA format for inhibitor screening.
  • Includes biotinylated TL1A and recombinant DR3 protein.
  • Provides a positive control neutralizing antibody and assay reagents.
  • Designed for 96-well screening and QC workflows.
  • Intended species: human.
  • Store unopened at 2-8 °C and follow component storage after opening.
  • No sensitivity or dynamic range specified on the product page.