Learn More
Enzo Life Sciences TL1A (human) monoclonal antibody (L4G9) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804832C100
Encompass_Preferred
Clone: L4G9. Isotype: IgG1. Immunogen: Recombinant human TL1A extracellular domain. Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human. Applications: ELISA, Flow Cytometry. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9992