Learn More
Enzo Life Sciences TL1A monoclonal antibody (Laety-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804859C100
Encompass_Preferred
Clone: Laety-1. Isotype: IgG2a. Immunogen: Recombinant human TL1A (ALX-522-084). Source: Purified from concentrated hybridoma tissue cell culture supernatant. Species reactivity: Human, Mouse. Applications: IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-0006