Learn More
Enzo Life Sciences TL1A (mouse) monoclonal antibody (Tingo-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804855C100
Encompass_Preferred
Clone: Tingo-1. Isotype: IgG1. Immunogen: Recombinant mouse TL1A extracellular domain (aa 94-252) (ALX-522-105). Source: Purified from concentrated hybridoma tissue cell culture supernatant. Species reactivity: Mouse. Applications: ELISA, IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-0003