Learn More
Enzo Life Sciences TL1A polyclonal antibody (AT127) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210952C100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant mouse TL1A (ALX-522-105). Source: Purified from rabbit serum. Species reactivity: Human, Mouse. Applications: ELISA, WB. Recommended Dilutions: Western Blot (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-8193