Learn More
BPS Bioscience Inc TL1A-Responsive Luciferase Reporter Jurkat Cell Line, 2 vials, Firefly Luciferase, TL1A and TNF-α stimulation, Dry ice shipping, Liquid nitrogen long-term storage
SDP
Catalog No. NC2585245
Encompass
The TL1A-Responsive Luciferase Reporter Jurkat Cell Line is a specialized tool designed for research in the NF-κB signaling pathway. This Jurkat cell line expresses human DR3 and utilizes firefly luciferase as a reporter gene to measure activity upon stimulation with TL1A or TNF-α. The absence of Mycoplasma species has been validated in this cell line, ensuring reliable results for research applications.
  • Product designed for monitoring NF-κB signaling pathway
  • Expresses human DR3 (death receptor 3)
  • Features firefly luciferase under NF-κB response element control
  • Enables measurement of soluble TL1A activity
  • Allows screening of anti-TL1A antibodies
  • Screened for Mycoplasma contamination
  • Supplied with 2 vials
  • Stored in liquid nitrogen for long-term preservation
  • Shipped on dry ice
  • Intended for research use only
Catalog No. NC2585245 Supplier BPS Bioscience Inc Supplier No. 78811
Add to Cart
Add to Cart
Request a Quote
The TL1A-Responsive Luciferase Reporter Jurkat Cell Line is a specialized tool designed for research in the NF-κB signaling pathway. This Jurkat cell line expresses human DR3 and utilizes firefly luciferase as a reporter gene to measure activity upon stimulation with TL1A or TNF-α. The absence of Mycoplasma species has been validated in this cell line, ensuring reliable results for research applications.
  • Product designed for monitoring NF-κB signaling pathway
  • Expresses human DR3 (death receptor 3)
  • Features firefly luciferase under NF-κB response element control
  • Enables measurement of soluble TL1A activity
  • Allows screening of anti-TL1A antibodies
  • Screened for Mycoplasma contamination
  • Supplied with 2 vials
  • Stored in liquid nitrogen for long-term preservation
  • Shipped on dry ice
  • Intended for research use only