Learn More
Enzo Life Sciences TL1A (soluble) (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX522084C010
Encompass_Preferred
Alternative name: TNFSF 15, VEGI. Concentration: 0.1mg/ml after reconstitution. Purity: ≥85% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in E. coli. The extracellular domain of human TL1A (aa 93-251) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG-tag. Specificity: Binds human DR3 and DcR3. Long Term Storage: -20°C. Applications: ELISA, Flow Cytometry .
Catalog No. 50-200-9409