Learn More
Invivogen TL8-506, 500 ug
SDP
Catalog No. NC1203873
Encompass
TL8-506 is a benzoazepine compound, analog of the Toll-like receptor 8 (TLR8) agonist VTX-2337. TL8-506 activates TLR8 more potently than the imidazoquinoline R848 (TLR7/8) and the thiazoquinolone CL075 (TLR8/7).
Catalog No. NC1203873 Supplier Invivogen Supplier No. TLRLTL8506
TL8-506 is a benzoazepine compound, analog of the Toll-like receptor 8 (TLR8) agonist VTX-2337. TL8-506 activates TLR8 more potently than the imidazoquinoline R848 (TLR7/8) and the thiazoquinolone CL075 (TLR8/7).