Learn More
Beckman Coulter TLA-110 Fixed-Angle Rotor, 110.000 rpm, 657.000 x g, 8 x 5.1 mL
Catalog No. NC2733102
Encompass
TLA-110 Rotor Package, Fixed Angle Rotor (28 deg Angle), Titanium.Major applications: Moderate volume differential sedimentation (pelleting) of subcellular organelles and viruses.For use in the Optima™ MAX-XP Benchtop Ultracentrifuge, Optima™ TLX personal micro-ultracentrifuges, and Optima MAX high-capacity personal micro-ultracentrifuges, and the discontinued TL-100 and Optima TL micro-ultracentrifuges.Note: May also be used in TL-100 with service installed spindle upgrade Part Number 360477.
Catalog No. NC2733102 Supplier Beckman Coulter Supplier No. 366735
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
TLA-110 Rotor Package, Fixed Angle Rotor (28 deg Angle), Titanium.Major applications: Moderate volume differential sedimentation (pelleting) of subcellular organelles and viruses.For use in the Optima™ MAX-XP Benchtop Ultracentrifuge, Optima™ TLX personal micro-ultracentrifuges, and Optima MAX high-capacity personal micro-ultracentrifuges, and the discontinued TL-100 and Optima TL micro-ultracentrifuges.Note: May also be used in TL-100 with service installed spindle upgrade Part Number 360477.