Learn More
Beckman Coulter TLA-55 Fixed-Angle Rotor Package for Optima MAX-XP and Optima TLX Ultracentrifuges - 12 x 1.5 mL, 55,000 rpm, 186,000 xg, 45° aluminum
Catalog No. NC1993025
Encompass
The TLA-55 Fixed-Angle Rotor Package is an aluminum fixed-angle rotor designed for high-speed ultracentrifugation applications, including rapid pelleting of nucleic acid precipitates from 1.5 mL tubes. It supports high rotational speeds and is intended for use with compatible benchtop and personal ultracentrifuges.
  • Maximum speed: 55,000 rpm (186,000 xg).
  • Capacity: 12 x 1.5 mL tubes (11 x 38 mm tube dimension), nominal capacity 18 mL.
  • Rotor angle: 45 degrees.
  • Material: aluminum construction for high-strength performance.
  • Suitable for rapid pelleting of nucleic acid precipitates and other small-volume preparations.
  • Compatible with selected benchtop and personal ultracentrifuge models; check equipment compatibility before use.
  • BioSafe lids supported where applicable.
This item is not returnable. View return policy
Catalog No. NC1993025 Supplier Beckman Coulter Supplier No. 366725
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
The TLA-55 Fixed-Angle Rotor Package is an aluminum fixed-angle rotor designed for high-speed ultracentrifugation applications, including rapid pelleting of nucleic acid precipitates from 1.5 mL tubes. It supports high rotational speeds and is intended for use with compatible benchtop and personal ultracentrifuges.
  • Maximum speed: 55,000 rpm (186,000 xg).
  • Capacity: 12 x 1.5 mL tubes (11 x 38 mm tube dimension), nominal capacity 18 mL.
  • Rotor angle: 45 degrees.
  • Material: aluminum construction for high-strength performance.
  • Suitable for rapid pelleting of nucleic acid precipitates and other small-volume preparations.
  • Compatible with selected benchtop and personal ultracentrifuge models; check equipment compatibility before use.
  • BioSafe lids supported where applicable.