Learn More
Medchemexpress LLC TLB 150 benzoate | 1208070-53-4 | 98.3% | 497.93 g·mol⁻¹ | C27H20ClN5O3 | 10 MG
SDP
Catalog No. 5000298556
Encompass_Preferred
TLB 150 benzoate (RAD150) is a research-stage androgen receptor modulator reported to have an IC50 of 0.13 μM; it is supplied as a benzoate ester for in vitro and preclinical research.
  • Reported androgen receptor activity (IC50 0.13 μM).
  • High purity suitable for laboratory assays (98.3%).
  • Molecular weight 497.93 g·mol⁻¹.
  • Provided in small research quantities, supporting dose-ranging studies.
  • Intended for research use in vitro and preclinical experiments.
Catalog No. 50-002-98556 Supplier Medchemexpress LLC Supplier No. HY4550910MG
TLB 150 benzoate (RAD150) is a research-stage androgen receptor modulator reported to have an IC50 of 0.13 μM; it is supplied as a benzoate ester for in vitro and preclinical research.
  • Reported androgen receptor activity (IC50 0.13 μM).
  • High purity suitable for laboratory assays (98.3%).
  • Molecular weight 497.93 g·mol⁻¹.
  • Provided in small research quantities, supporting dose-ranging studies.
  • Intended for research use in vitro and preclinical experiments.